Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137159.4 | NP_611003.2 | 1 | 980 | 130 | |
GenBank mRNA | BT030785.1 | - | 1 | 985 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |