Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202087.1 | NP_001189016.1 | 2 | 2236 | 64 | |
NM_138243.4 | NP_612087.1 | 2 | 2274 | 64 | ||
GenBank mRNA | AY047571.1 | - | 2 | 2275 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |