Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142349.3 | NP_650606.2 | 2 | 1042 | 75 | |
GenBank mRNA | BT001478.1 | - | 2 | 1022 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |