Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssdp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275769.1 | NP_001262698.1 | 2 | 3074 | 109 | |
NM_142431.1 | NP_650688.1 | 2 | 3074 | 109 | ||
NM_169794.2 | NP_732304.1 | 2 | 2569 | 109 | ||
NM_169795.1 | NP_732305.1 | 2 | 3062 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |