Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNUTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014453.2 | NP_001014453.1 | 5 | 3532 | 53 | |
NM_001272921.1 | NP_001259850.1 | 6 | 2744 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |