Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggamma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078938.3 | NP_523662.1 | 2 | 269 | 69 | |
NM_165631.2 | NP_724718.1 | 2 | 331 | 69 | ||
NM_165632.2 | NP_724719.1 | 2 | 522 | 69 | ||
GenBank mRNA | AY118489.1 | - | 2 | 502 | 69 | |
M85042.1 | - | 2 | 316 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |