Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258527.1 | NP_001245456.1 | 2 | 766 | 56 | |
NM_001272186.1 | NP_001259115.1 | 3 | 799 | 56 | ||
NM_166866.2 | NP_726710.1 | 2 | 920 | 56 | ||
NM_166867.2 | NP_726711.1 | 2 | 1243 | 56 | ||
NM_166868.1 | NP_726712.1 | 2 | 1039 | 56 | ||
GenBank mRNA | AY075393.1 | - | 2 | 1243 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |