Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS15Aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169267.2 | NP_001162738.1 | 3 | 476 | 74 | |
NM_079970.6 | NP_524709.1 | 2 | 543 | 74 | ||
NM_167360.3 | NP_727690.1 | 3 | 480 | 74 | ||
NM_167362.3 | NP_727692.1 | 3 | 539 | 74 | ||
NM_167363.3 | NP_727693.1 | 2 | 543 | 74 | ||
GenBank mRNA | AY084182.1 | - | 3 | 488 | 74 | |
Z21673.1 | - | 465 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |