Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258821.2 | NP_001245750.1 | 3 | 2922 | 62 | |
NM_001258822.2 | NP_001245751.1 | 2 | 3058 | 62 | ||
NM_001258823.2 | NP_001245752.1 | 2 | 3244 | 62 | ||
NM_001298511.1 | NP_001285440.1 | 2 | 2892 | 62 | ||
NM_001298512.1 | NP_001285441.1 | 2 | 3234 | 62 | ||
NM_058104.5 | NP_477452.3 | 2 | 3237 | 62 | ||
NM_167651.2 | NP_728249.1 | 2 | 2842 | 62 | ||
NM_167652.2 | NP_728250.1 | 2 | 2844 | 62 | ||
NM_167653.3 | NP_728251.1 | 2 | 3054 | 62 | ||
NM_167654.2 | NP_728252.1 | 2 | 2773 | 62 | ||
NM_167655.2 | NP_728253.1 | 2 | 2841 | 62 | ||
NM_167656.2 | NP_728254.1 | 2 | 2839 | 62 | ||
GenBank mRNA | AY122115.1 | - | 2 | 3239 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |