Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079228.3 | NP_523952.2 | 2 | 305 | 78 | |
NM_168192.3 | NP_729218.1 | 3 | 339 | 78 | ||
GenBank mRNA | AY070704.1 | - | 3 | 331 | 78 | |
U04823.1 | - | 3 | 309 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |