Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VepD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259530.2 | NP_001246459.1 | 2 | 242 | 96 | |
NM_001299791.1 | NP_001286720.1 | 2 | 366 | 96 | ||
NM_176245.4 | NP_788425.1 | 2 | 242 | 96 | ||
GenBank mRNA | AJ276848.1 | - | 2 | 240 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |