Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubi-p63E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274454.1 | NP_001261383.1 | 2 | 2450 | 106 | |
NM_079185.4 | NP_523909.2 | 3 | 2585 | 106 | ||
NM_206272.1 | NP_995994.1 | 2 | 2536 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |