Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014589.4 | NP_001014589.1 | 8 - 9 | 1243 | 71 | |
NM_001038939.2 | NP_001034028.2 | 7 - 8 | 1170 | 71 | ||
GenBank mRNA | BT031129.1 | - | 8 - 9 | 1209 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |