Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CanA-14F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258788.2 | NP_001245717.1 | 2 | 2703 | 128 | |
NM_001272693.2 | NP_001259622.1 | 2 | 2376 | 128 | ||
NM_167523.3 | NP_727985.2 | 2 | 2579 | 128 | ||
NM_167524.3 | NP_727986.2 | 2 | 2568 | 128 | ||
GenBank mRNA | BT004864.1 | - | 2 | 2534 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |