Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103385.2 | NP_001096855.1 | 11 - 12 | 3937 | 64 | |
NM_001103386.2 | NP_001096856.1 | 11 - 12 | 3937 | 64 | ||
NM_001297795.1 | NP_001284724.1 | 11 - 12 | 3938 | 64 | ||
NM_001297796.1 | NP_001284725.1 | 11 - 12 | 3944 | 64 | ||
GenBank mRNA | BT024449.1 | - | 11 - 12 | 3899 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |