Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042986.3 | NP_001036451.1 | 16 - 17 | 4695 | 150 | |
NM_001042987.3 | NP_001036452.1 | 16 - 17 | 4723 | 150 | ||
NM_001169581.2 | NP_001163052.1 | 17 - 18 | 4616 | 150 | ||
NM_001169582.2 | NP_001163053.1 | 17 - 18 | 4651 | 150 | ||
NM_001169583.2 | NP_001163054.1 | 17 - 18 | 4623 | 150 | ||
NM_001273774.1 | NP_001260703.1 | 15 - 16 | 4363 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |