Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheA86a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032003.2 | NP_001027174.1 | 1 - 2 | 141 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |