Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pHCl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038935.3 | NP_001034024.2 | 7 - 8 | 1676 | 77 | |
NM_001038936.4 | NP_001034025.2 | 7 - 8 | 1673 | 77 | ||
NM_001038937.2 | NP_001034026.1 | 7 - 8 | 1676 | 77 | ||
NM_001169979.1 | NP_001163450.1 | 7 - 8 | 1676 | 77 | ||
NM_001259852.1 | NP_001246781.1 | 7 - 8 | 1681 | 77 | ||
NM_001274965.1 | NP_001261894.1 | 6 - 7 | 1660 | 77 | ||
NM_001274968.1 | NP_001261897.1 | 6 - 7 | 1655 | 77 | ||
NM_001274969.1 | NP_001261898.1 | 7 - 8 | 1676 | 77 | ||
NM_001274971.1 | NP_001261900.1 | 6 - 7 | 1655 | 77 | ||
NM_001370024.1 | NP_001356981.1 | 6 - 7 | 1660 | 77 | ||
NM_001370025.1 | NP_001356910.1 | 7 - 8 | 1681 | 77 | ||
GenBank mRNA | BT012472.1 | - | 7 - 8 | 1677 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |