Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pHCl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038935.3 | NP_001034024.2 | 11 - 12 | 2178 | 91 | |
NM_001038936.4 | NP_001034025.2 | 11 - 12 | 2175 | 91 | ||
NM_001169979.1 | NP_001163450.1 | 11 - 12 | 2178 | 91 | ||
NM_001259853.1 | NP_001246782.1 | 10 - 11 | 2108 | 91 | ||
NM_001274966.1 | NP_001261895.1 | 9 - 10 | 2087 | 91 | ||
NM_001274970.1 | NP_001261899.1 | 9 - 10 | 2082 | 91 | ||
NM_001370025.1 | NP_001356910.1 | 11 - 12 | 2183 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |