Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258761.3 | NP_001245690.1 | 2 - 3 | 2406 | 57 | |
NM_001258762.3 | NP_001245691.1 | 2 - 3 | 2406 | 57 | ||
NM_001272642.2 | NP_001259571.2 | 2 - 3 | 1879 | 57 | ||
NM_001370020.1 | NP_001356908.1 | 2 - 3 | 1879 | 57 | ||
NM_132794.5 | NP_573022.2 | 2 - 3 | 2406 | 57 | ||
NM_167441.5 | NP_727848.3 | 2 - 3 | 2406 | 57 | ||
GenBank mRNA | BT010109.1 | - | 3 - 4 | 1504 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |