Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubi-p63E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274454.1 | NP_001261383.1 | 2 | 2407 | 87 | |
NM_079185.4 | NP_523909.2 | 3 | 2542 | 87 | ||
NM_168043.3 | NP_728908.1 | 2 | 2476 | 87 | ||
NM_206272.1 | NP_995994.1 | 2 | 2493 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |