Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103971.3 | NP_001097441.1 | 4 - 5 | 718 | 134 | |
NM_001103972.3 | NP_001097442.2 | 4 - 5 | 718 | 134 | ||
NM_001144276.2 | NP_001137748.1 | 4 - 5 | 732 | 134 | ||
NM_001144277.2 | NP_001137749.1 | 4 - 5 | 732 | 134 | ||
NM_001299886.1 | NP_001286815.1 | 4 - 5 | 718 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |