Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104200.3 | NP_001097670.1 | 1 - 2 | 343 | 126 | |
GenBank mRNA | AB232987.1 | - | 1 - 2 | 153 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |