Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vtd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015132.2 | NP_001015132.1 | 7 - 8 | 2108 | 140 | |
NM_001300711.1 | NP_001287640.1 | 8 - 9 | 2336 | 140 | ||
GenBank mRNA | AF109926.1 | - | 7 - 8 | 2058 | 140 | |
BT100037.1 | - | 7 - 8 | 2055 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |