Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Set1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015221.3 | NP_001015221.1 | 2 - 3 | 354 | 61 | |
NM_001015222.3 | NP_001015222.1 | 2 - 3 | 537 | 61 | ||
NM_001110936.3 | NP_001104406.1 | 2 - 3 | 546 | 61 | ||
NM_001170375.2 | NP_001163846.1 | 3 - 4 | 362 | 61 | ||
NM_001170376.2 | NP_001163847.1 | 2 - 3 | 377 | 61 | ||
NM_001170377.1 | NP_001163848.1 | 2 - 3 | 341 | 61 | ||
NM_001170378.1 | NP_001163849.1 | 2 - 3 | 330 | 61 | ||
NM_001170379.1 | NP_001163850.1 | 1 - 2 | 327 | 61 | ||
NM_001170380.2 | NP_001163851.1 | 2 - 3 | 368 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |