Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015363.4 | NP_001015363.3 | 3 - 4 | 506 | 65 | |
NM_001170373.2 | NP_001163844.1 | 3 - 4 | 560 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |