Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015499.4 | NP_001015499.4 | 5 - 6 | 1160 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |