Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015499.4 | NP_001015499.4 | 7 - 8 | 1842 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |