Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038721.2 | NP_001033810.1 | 5 - 6 | 2332 | 109 | |
NM_001038723.2 | NP_001033812.1 | 4 - 5 | 2268 | 109 | ||
NM_001038725.2 | NP_001033814.1 | 5 - 6 | 2332 | 109 | ||
GenBank mRNA | AY118401.1 | - | 5 - 6 | 2326 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |