Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042950.3 | NP_001036415.2 | 5 - 6 | 904 | 82 | |
NM_001042951.2 | NP_001036416.1 | 5 - 6 | 904 | 82 | ||
NM_001042952.2 | NP_001036417.1 | 4 - 5 | 956 | 82 | ||
NM_001273766.1 | NP_001260695.1 | 5 - 6 | 904 | 82 | ||
GenBank mRNA | AY118388.1 | - | 4 - 5 | 964 | 82 | |
BT024228.1 | - | 5 - 6 | 861 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |