Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp54A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043089.3 | NP_001036554.1 | 1 | 144 | 86 | |
GenBank mRNA | BT053719.1 | - | 1 | 131 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |