Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014642.5 | NP_001014642.4 | 5 - 6 | 1951 | 87 | |
NM_001043267.3 | NP_001036732.1 | 5 - 6 | 1951 | 87 | ||
NM_001260277.2 | NP_001247206.1 | 4 - 5 | 981 | 87 | ||
NM_001260278.2 | NP_001247207.1 | 5 - 6 | 1951 | 87 | ||
NM_001260279.2 | NP_001247208.1 | 5 - 6 | 1951 | 87 | ||
NM_001260280.2 | NP_001247209.1 | 4 - 5 | 995 | 87 | ||
NM_001260281.2 | NP_001247210.1 | 4 - 5 | 1013 | 87 | ||
NM_001275827.2 | NP_001262756.1 | 5 - 6 | 1951 | 87 | ||
NM_001300494.1 | NP_001287423.1 | 4 - 5 | 995 | 87 | ||
NM_001300495.1 | NP_001287424.1 | 5 - 6 | 1951 | 87 | ||
GenBank mRNA | BT001445.1 | - | 5 - 6 | 1914 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |