Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258680.2 | NP_001245609.1 | 5 - 6 | 919 | 80 | |
NM_001258681.2 | NP_001245610.1 | 2 - 3 | 661 | 80 | ||
NM_001258682.2 | NP_001245611.1 | 5 - 6 | 859 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |