Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaF-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103489.2 | NP_001096959.1 | 1 | 472 | 84 | |
GenBank mRNA | BT030281.1 | - | 1 | 472 | 84 | |
KX532126.1 | - | 1 | 353 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |