Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 76 - 77 | 16364 | 65 | |
XM_005165511.4 | XP_005165568.1 | 76 - 77 | 16483 | 65 | ||
XM_009301753.3 | XP_009300028.1 | 77 - 78 | 16505 | 65 | ||
GenBank mRNA | AY540131.1 | - | 76 - 77 | 16368 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |