Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002125.1 | NP_001002125.1 | 18 - 19 | 1476 | 85 | |
XM_005160662.4 | XP_005160719.1 | 19 - 20 | 1591 | 85 | ||
XM_005160663.4 | XP_005160720.1 | 18 - 19 | 1501 | 85 | ||
XM_005160664.4 | XP_005160721.1 | 17 - 18 | 1438 | 85 | ||
XM_017352461.2 | XP_017207950.1 | 17 - 18 | 1432 | 85 | ||
XM_021468390.1 | XP_021324065.1 | 20 - 21 | 1666 | 85 | ||
XM_021468391.1 | XP_021324066.1 | 20 - 21 | 1648 | 85 | ||
XM_021468392.1 | XP_021324067.1 | 19 - 20 | 1603 | 85 | ||
XM_021468393.1 | XP_021324068.1 | 18 - 19 | 1522 | 85 | ||
GenBank mRNA | BC071450.1 | - | 18 - 19 | 1476 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |