Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm5bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002166.2 | NP_001002166.2 | 3 - 4 | 531 | 98 | |
XM_005167024.4 | XP_005167081.1 | 3 - 4 | 1152 | 98 | ||
XM_009303989.3 | XP_009302264.1 | 3 - 4 | 1152 | 98 | ||
GenBank mRNA | BC071280.1 | - | 3 - 4 | 464 | 98 | |
EH449360.1 | - | 3 - 4 | 541 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |