Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camk2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003602.2 | NP_001003602.1 | 16 - 17 | 1409 | 73 | |
NM_001114708.2 | NP_001108180.1 | 17 - 18 | 1454 | 73 | ||
XM_005166561.4 | XP_005166618.1 | 16 - 17 | 1445 | 73 | ||
XM_009303481.3 | XP_009301756.1 | 18 - 19 | 1792 | 73 | ||
XM_009303482.3 | XP_009301757.1 | 18 - 19 | 1793 | 73 | ||
XM_009303484.3 | XP_009301759.1 | 17 - 18 | 1747 | 73 | ||
XM_009303485.3 | XP_009301760.1 | 17 - 18 | 1490 | 73 | ||
XM_021477468.1 | XP_021333143.1 | 18 - 19 | 1551 | 73 | ||
GenBank mRNA | BC077143.1 | - | 16 - 17 | 1377 | 73 | |
EE714463.1 | - | 4 - 5 | 268 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |