Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nicn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003610.2 | NP_001003610.1 | 4 - 5 | 579 | 69 | |
XM_009296489.3 | XP_009294764.1 | 4 - 5 | 733 | 69 | ||
GenBank mRNA | BC077148.1 | - | 4 - 5 | 583 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |