Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003640.2 | NP_001003640.1 | 3 - 4 | 533 | 63 | |
GenBank mRNA | BC078304.1 | - | 3 - 4 | 537 | 63 | |
KC136847.1 | - | 2 - 3 | 326 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |