Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003993.1 | NP_001003993.1 | 4 - 5 | 388 | 100 | |
XM_005168682.4 | XP_005168739.1 | 3 - 4 | 285 | 100 | ||
XM_005168683.4 | XP_005168740.1 | 4 - 5 | 469 | 100 | ||
XM_009302056.3 | XP_009300331.1 | 4 - 5 | 385 | 100 | ||
XM_017356070.2 | XP_017211559.1 | 5 - 6 | 598 | 100 | ||
GenBank mRNA | BC080261.1 | - | 4 - 5 | 388 | 100 | |
BC163982.1 | - | 2 - 3 | 245 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |