Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004657.1 | NP_001004657.1 | 6 - 7 | 724 | 102 | |
XM_005163809.4 | XP_005163866.1 | 6 - 7 | 760 | 102 | ||
XM_021470416.1 | XP_021326091.1 | 6 - 7 | 684 | 102 | ||
GenBank mRNA | BC081519.1 | - | 6 - 7 | 724 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |