Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004657.1 | NP_001004657.1 | 7 - 8 | 861 | 137 | |
XM_005163809.4 | XP_005163866.1 | 7 - 8 | 897 | 137 | ||
XM_021470416.1 | XP_021326091.1 | 7 - 8 | 821 | 137 | ||
GenBank mRNA | BC081519.1 | - | 7 - 8 | 861 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |