Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007219.1 | NP_001007220.1 | 5 - 6 | 901 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Danio rerio Inventoried Probe spans exons |