Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013292.2 | NP_001013310.1 | 4 - 5 | 1147 | 66 | |
GenBank mRNA | BC090267.1 | - | 4 - 5 | 1151 | 66 | |
BC129289.1 | - | 4 - 5 | 1121 | 66 | ||
BC165795.1 | - | 3 - 4 | 1019 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |