Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017637.2 | NP_001017637.1 | 5 - 6 | 599 | 88 | |
GenBank mRNA | BC092813.1 | - | 5 - 6 | 540 | 88 | |
EH538457.1 | - | 5 - 6 | 622 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |