Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017656.1 | NP_001017656.1 | 6 - 7 | 740 | 82 | |
XM_005162505.3 | XP_005162562.1 | 6 - 7 | 760 | 82 | ||
XM_021470163.1 | XP_021325838.1 | 7 - 8 | 845 | 82 | ||
GenBank mRNA | BC093175.1 | - | 6 - 7 | 740 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |