Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007323.2 | NP_001007324.2 | 5 - 6 | 1029 | 71 | |
XM_005165077.3 | XP_005165134.1 | 5 - 6 | 1020 | 71 | ||
GenBank mRNA | BC085627.1 | - | 5 - 6 | 774 | 71 | |
BC095654.1 | - | 5 - 6 | 980 | 71 | ||
BC164655.1 | - | 5 - 6 | 754 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |