Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039719.2 | NP_001034808.2 | 6 - 7 | 1286 | 62 | |
GenBank mRNA | BC076205.1 | - | 6 - 7 | 1240 | 62 | |
BC093187.1 | - | 6 - 7 | 1242 | 62 | ||
BC114247.1 | - | 6 - 7 | 1271 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |