Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042778.2 | NP_001036243.2 | 5 - 6 | 890 | 91 | |
GenBank mRNA | BC134103.1 | - | 5 - 6 | 890 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |